Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ ANA Screen ELISA Kit

Catalog No. 89024480 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
96 Assays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-024-480 96 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-024-480 Supplier Abnova™ Supplier No. KA0939
Add to Cart
Add to Cart

Includes

  • Microwells coated with nuclear antigens, 96 wells
  • Sample Diluent (ready to use), 22mL
  • Calibrator (ready to use), 1.5mL
  • Positive Control (ready to use), 1.5mL
  • Negative Control (ready to use), 1.5mL
  • Enzyme conjugate (ready to use), 12mL
  • TMB Substrate (ready to use), 12mL
  • Stop Solution (ready to use), 12mL
  • Wash concentrate 20X, 25mL

An enzyme-linked immunosorbent assay (ELISA) for the detection of IgG class antibodies to ANA in human serum or plasma.

Specifications

Conjugate HRP
Cross Reactivity Human
Description ANA Screen ELISA Kit is an enzyme-linked immunosorbent assay (ELISA) for the detection of IgG class antibodies to ANA in human serum or plasma.
Detection Method Colorimetric
Includes 12 x 8 x 1 Microwell coated with nuclear Antigens, 22mL sample diluent, 1.5mL calibrator, 1.5mL positive control, 1.5mL negative control, 12mL Enzyme Conjugate, 12mL TMB Substrate, 12mL stop solution, 25mL wash concentrate 20X
Quantity 96 Assays
Sample Type Plasma, Serum
Sample Volume 10 μL
Content And Storage Store the kit at 4°C
For Use With (Application) Qualitative
Product Type ANA Screen ELISA Kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.