Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ ANA Screen ELISA Kit

Catalog No. 89027219 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
96 Assays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-027-219 96 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-027-219 Supplier Abnova™ Supplier No. KA1081
Add to Cart
Add to Cart

Qualitative, intended to screen for the presence of antinuclear antibodies in Human Serum or Plasma

Specifications

Conjugate HRP
Description ANA Screen ELISA Kit is a assay intended to screen for the presence of antinuclear antibodies (ANAs) in human serum or plasma.
Detection Method Colorimetric
Includes 2 Vials, 1,5mL each Anti-ANA controls, 1 vial 20mL Sample Buffer, 1 vial 15mL Enzyme Conjugate solution, 1 vial 15 Ml TMB Substrate solution, 1 vial 15mL stop solution, 1 plate 96-well plate, 1 vial 20mL wash solution
Quantity 96 Assays
Sample Type Plasma, Serum
Sample Volume 10 μL
Content And Storage Store the kit at 4°C
For Use With (Application) Qualitative
Product Type ANA Screen ELISA Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.