Learn More
Specifications
Specifications
| Conjugate | HRP |
| Description | ANCA-C (PR3) is an indirect solid phase ELISA for the quantitative measurement of IgG class autoantibodies against proteinase 3 (PR3) in human serum or plasma. |
| Detection Method | Colorimetric |
| Includes | 6 Vials, 1.5mL each combined calibrator with IgG class ANCA-C (PR3) Antibodies, 2 vials, 1.5mL each ANCA-C (PR3) controls, 1 vial, 20mL Sample Buffer, 1 vial 15mL Enzyme Conjugate solution, 1 vial 15mL TMB Substrate solution, 1 plate 96-well plate, 1 vial 15mL stop solution, 1 vial 20mL wash solution |
| Quantity | 96 Assays |
| Sample Type | Plasma, Serum |
| Sample Volume | 10 μL |
| Content And Storage | Store the kit at 4°C |
| For Use With (Application) | Quantification |
| Product Type | ANCA-C (PR3) ELISA Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.