Learn More
Description
Includes
- One divisible microplate consisting of 12 modules of 8 wells each, coated with highly purified antigens: PR3, MPO, BPI, Elastase, Cathepsin G, Lysozyme and
- Lactoferrin. Ready to use.
- 3 vials, 1.5mL each ANCA Controls in a serum/buffer matrix (PBS, BSA, NaN3 <0.1% (w/w)) Negative Control (A), Cut-Off control (B) and Positive Control (C), for the respective concentrations see the enclosed package insert. Ready to use
- 1 vial, 20mL Sample buffer (Tris, NaN3 <0.1% (w/w)), yellow, concentrate (5x)
- 1 vial, 15mL Enzyme conjugate solution (PBS, Proclin 300 <0.5% (v/v)), (light red) containing polyclonal rabbit anti-human IgG; labelled with horseradish peroxidase. Ready to use
- 1 vial, 15mL TMB substrate solution. Ready to use
- 1 vial, 15mL Stop solution (1M hydrochloric acid). Ready to use
- 1 vial, 20mL Wash solution (PBS, NaN3 <0.1% (w/w)), concentrate (50x)
Specifications
Specifications
| Conjugate | HRP |
| Description | ANCA Profile ELISA Kit is an indirect solid phase ELISA for the determination of anti-neutrophil cytoplasmic antibodies (ANCA) directed against PR3, MPO, BPI, Elastase, Cathepsin G, Lysozyme and Lactoferrin in human serum or plasma. |
| Detection Method | Colorimetric |
| Includes | 3 Vials 1.5mL each ANCA controls, 1 vial 20mL Sample Buffer, 1 vial 15mL Enzyme Conjugate solution, 1 vial 15mL TMB Substrate solution, 1 vial 15mL wash solution stop solution, 1 plate 96-well plate, 1 vial 20mL wash solution |
| Quantity | 96 Tests |
| Sample Type | Plasma, Serum |
| Sample Volume | 10 μL |
| Content And Storage | Store the kit at 4°C |
| For Use With (Application) | Qualitative |
| Product Type | ANCA Profile ELISA Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.