Learn More
Specifications
Specifications
| Conjugate | HRP |
| Description | ANCAbScreen ELISA Kit is a assay intended to screen for the presence of IgG class autoantibodies against PR3 and MPO in human serum or plasma. |
| Detection Method | Colorimetric |
| Includes | 3 Vials 1.5mL each Anti-ANA controls, 1 vial 20mL Sample Buffer, 1 vial, 15mL Enzyme Conjugate solution, 1 vial 15mL TMB Substrate solution, 1 vial 15mL stop solution, 1 plate 96-well plate, 1 vial 20mL wash solution |
| Quantity | 96 Assays |
| Sample Type | Plasma, Serum |
| Sample Volume | 10 μL |
| Content And Storage | Store the kit at 4°C |
| For Use With (Application) | Qualitative |
| Product Type | ANCA screen ELISA Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.