Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Antifade Solution

Catalog No. 89028831 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 x 0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-831 2 x 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-831 Supplier Abnova™ Supplier No. U0032
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For use with In situ Hybridization.

Antifade Solution is comprised of Antifade and glycerol in an aqueous buffer solution.

Specifications

Content And Storage Store at 4°C
Description Antifade Solution is comprised of Antifade and glycerol in an aqueous buffer solution.
For Use With (Application) Fluorescent In Situ Hybridization (Cell)
Product Type Antifade Solution
Quantity 2 x 0.5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.