Learn More
Specifications
Specifications
| Conjugate | HRP |
| Cross Reactivity | Rat |
| Description | Apcs (Rat) is a highly sensitive two-site enzyme-linked immunoassay for the quantitative determination of Apcs in rat biological samples. |
| Detection Method | Colorimetric |
| Includes | 50mL Diluent concentrate, 50mL wash solution concentrate, 150μL Enzyme-Antibody Conjugate concentrate, 12mL TMB Substrate solution, 12mL stop solution, 96-well microtiter plate, 1 vial calibrator |
| Quantity | 1 Kit |
| Sample Type | Plasma, Serum |
| Sample Volume | 100 μL |
| Content And Storage | Store the kit at 4°C |
| For Use With (Application) | Quantification |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.