Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ APOA2 (Human) ELISA Kit

Catalog No. 89100371 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Type:
CCS, Milk, Plasma, Serum, Urine
CCS, Plasma, Serum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
89-100-371 CCS, Plasma, Serum
89-022-888 CCS, Milk, Plasma, Serum, Urine
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-100-371 Supplier Abnova™ Supplier No. KA2113
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • Human Apo A-II Microplate: A 96-well polystyrene microplate (12 strips of 8 wells) coated with a monoclonal antibody against human Apo A-II
  • Sealing Tapes: Each kit contains 3 pre-cut, pressure-sensitive sealing tapes that can be cut to fit the format of the individual assay
  • Human Apo A-II Standard: Human Apo A-II in a buffered protein base (μg, lyophilized)
  • Biotinylated Apo A-II Antibody (25x): A 25-fold concentrated biotinylated polyclonal antibody against Apo A-II (280μL)
  • EIA Diluent Concentrate (10x): A 10-fold concentrated buffered protein base (30mL)
  • Wash Buffer Concentrate (20x): A 20-fold concentrated buffered surfactant (30mL, 2 bottles)
  • Streptavidin-Peroxidase Conjugate (SP Conjugate): A 100-fold concentrate (80μL)
  • Chromogen Substrate: A ready-to-use stabilized peroxidase chromogen substrate tetramethylbenzidine (8mL)

Sandwich Enzyme Immunoassay for the quantitative measurement of Human APOA2

Specifications

Cross Reactivity Human
Description APOA2 (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human APOA2.
Detection Method Colorimetric
Quantity 1 Kit
Sample Type CCS, Plasma, Serum
Sample Volume 50 μL
Content And Storage Store components of the kit at 4°C or -20°C as described in the protocol.
For Use With (Application) Quantification
Product Type APOA2 (Human) ELISA kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.