Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ APOB (Human) ELISA Kit

Catalog No. 89028162 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-162 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-162 Supplier Abnova™ Supplier No. KA1028
Add to Cart
Add to Cart

Includes

  • Human Apo B Microplate: A 96-well polystyrene microplate (12 strips of 8 wells) coated with a polyclonal antibody against human Apo B, 1 in no.
  • Pressure-sensitive sealing tapes that can be cut to fit the format of the individual assay, 3 in nos.
  • Human Apo B standard in a buffered protein base ( lyophilized), 0.75μg
  • Biotinylated Apo B Antibody (100x): A 100-fold concentrated biotinylated polyclonal antibody against Apo B, 80μL
  • EIA Diluent Concentrate (10x): A 10-fold concentrated buffered protein base, 30mL
  • Wash Buffer Concentrate (20x): A 20-fold concentrated buffered surfactant, 30mL, 2 bottles
  • Streptavidin-Peroxidase Conjugate (SP Conjugate): A 100-fold concentrate, 80μL
  • Ready-to-use stabilized peroxidase chromogen substrate tetramethylbenzidine, 8mL
  • Stop Solution: A 0.5 N hydroxychloric acid to stop the chromogen substrate reaction, 12mL

Sandwich Enzyme Immunoassay for the quantitative measurement of Human APOB in Plasma, Serum and cell culture samples

Specifications

Conjugate HRP
Cross Reactivity Human
Description APOB (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human APOB.
Detection Method Colorimetric
Includes 96-well Antibody coated microwell, 0.75ug Human apo B standard, 1 via Biotinylated apo B Antibody (100X), 30mL EIA diluent concentrate (10X), 30mL x 2 Wash Buffer 20X, 80 Streptavidin-Peroxidase Conjugate (SP Conjugate), 8mL chromogen Substrate, 12mL stop solution
Quantity 1 Kit
Sample Type Cell Culture Supernatant, Plasma, Serum
Sample Volume 50 μL
Content And Storage Store components of the kit at 4°C or -20°C as described in the protocol.
For Use With (Application) Quantification
Product Type APOB (Human) ELISA kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.