Learn More
Description
Includes
- Human Apo C-III Microplate: A 96-well polystyrene microplate (12 strips of 8 wells) coated with a polyclonal antibody against human Apo C-III - 1
- Sealing Tapes: pressure-sensitive sealing tapes that can be cut to fit the format of the individual assay- 3
- Human Apo C-III Standard: Human Apo C-III in a buffered protein base 4μg, lyophilized
- Biotinylated Apo C-III Antibody (100x): A 100-fold concentrated biotinylated polyclonal antibody against Apo C-III 80μL
- EIA Diluent Concentrate (10x): A 10-fold concentrated buffered protein base 30mL
- Wash Buffer Concentrate (20x): A 20-fold concentrated buffered surfactant 30mL , 2 bottles
- Streptavidin-Peroxidase Conjugate (SP Conjugate): A 100-fold concentrate 80μL
- Chromogen Substrate: A ready-to-use stabilized peroxidase chromogen substrate tetramethylbenzidine 8mL
- Stop Solution: A 0.5 N hydroxychloric acid to stop the chromogen substrate reaction 12mL
Specifications
Specifications
| Conjugate | Biotin |
| Cross Reactivity | Human |
| Description | APOC3 (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human APOC3. |
| Detection Method | Colorimetric |
| Includes | 96-well Antibody coated microwell, 10μg Human apoC-II standard, 1 via Biotinylated apo B Antibody (100X), 30mL EIA diluent concentrate (10X), 30mL Wash Buffer 20X, 80 Streptavidin-Peroxidase Conjugate (SP Conjugate), 8mL chromogen Substr |
| Quantity | 1 Kit |
| Sample Type | Cell Culture Supernatant, Milk, Plasma, Serum, Urine |
| Sample Volume | 50 μL |
| Content And Storage | Store SP Conjugate and Biotinylated Antibody at -20°C. Store Standard at 2-8°C before reconstituting with Diluent and at -20°C after reconstituting with Diluent. Store all other components at 4°C. |
| For Use With (Application) | Quantification |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.