Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ APOE (Human) ELISA Kit

Catalog No. 89028161 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-161 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-161 Supplier Abnova™ Supplier No. KA1031
Add to Cart
Add to Cart

Includes

  • Human Apo E Microplate: A 96-well polystyrene microplate (12 strips of 8 wells) coated with a polyclonal antibody against human ApoE, 1 in no.
  • Pressure-sensitive sealing tapes that can be cut to fit the format of the individual assay, 3 in nos.
  • Human Apo E standard in a buffered protein base, lyophilized, 1μg, 2 vials
  • Biotinylated Apo E antibody (100x): A 100-fold concentrated biotinylated polyclonal antibody against Apo E, 80μL
  • EIA Diluent Concentrate (10x): A 10-fold concentrated buffered protein base 30mL
  • Wash Buffer Concentrate (20x): A 20-fold concentrated buffered surfactant 30mL, 2 bottles
  • Streptavidin-Peroxidase Conjugate (SP Conjugate): A 100-fold concentrate 80μLl
  • A ready-to-use stabilized peroxidase chromogen substrate tetramethylbenzidine, 8mL
  • Stop Solution: 0.5N hydroxychloric acid to stop the chromogen substrate reaction 12mL

Sandwich Enzyme Immunoassay for the quantitative measurement of Human APOE in Plasma, Serum, Urine and cell culture samples

Specifications

Conjugate HRP
Cross Reactivity Human
Description APOE (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human APOE in plasma, serum, urine, and cell culture samples.
Detection Method Colorimetric
Includes 96-well Antibody coated microwell, 1μg x 2 vials Human Albumin standard (lyophilized), 80μL Biotinylated apo E Antibody (100X), 30mL EIA Diluent concentrate (10X), 30mL Wash Buffer 20X, 80μL Streptavid
Quantity 1 Kit
Sample Type Cell Culture Supernatant, Cerebrospinal Fluid, Plasma, Serum
Sample Volume 50 μL
Content And Storage Store components of the kit at 4°C or -20°C as described in the protocol.
For Use With (Application) Quantification
Product Type APOE (Human) ELISA kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.