Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ APOH (Human) ELISA Kit

Catalog No. 89028037 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
96 Assays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-037 96 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-037 Supplier Abnova™ Supplier No. KA0982
Add to Cart
Add to Cart

Designed for detection of human APOH

  • Sample Volume: 50μL
  • Sample Type:: Cell Culture Supernatant, Plasma, Saliva, Serum, Urine
  • Calibration Range: 0.625 to 40ng/mL
  • Detection Method:: Colorimetric method

Applications:

Quantification

Specifications

Conjugate Biotin
Cross Reactivity Human
Description APOH (Human) ELISA Kit is designed for detection of human APOH in plasma, serum, urine, and cell culture samples.
Detection Method Colorimetric
Includes 96-well Antibody coated microwell, 320ng Human Apo H standard, 1 vial Biotinylated Apo H Antibody (100X), 30mL mix diluent concentrate (10X), 30mL X 2 Wash Buffer 20X, 80 Streptavidin-Peroxidase Conjugate (SP Conjugate), 8mL chromogen Substrate, 12mL stop solution
Quantity 96 Assays
Sample Type Cell Culture Supernatant, Milk, Plasma, Saliva, Serum, Urine
Sample Volume 50 μL
Content And Storage Store components of the kit at 4°C or -20°C as described in the protocol.
For Use With (Application) Quantification
Product Type APOH (Human) ELISA Kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.