Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ ASCA IgG/IgA ELISA Kit

Catalog No. 89024562 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-024-562 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-024-562 Supplier Abnova™ Supplier No. KA1270
Add to Cart
Add to Cart

Indirect solid phase Enzyme Immunoassay for quantitative measurement of IgG and IgA class autoantibodies against mannan from Saccharomyces cerevisiae in Human Serum or Plasma

Specifications

Conjugate HRP
Cross Reactivity Human
Description ASCA ELISA Kit is an indirect solid phase enzyme immunoassay for the quantitative measurement of IgG and IgA class autoantibodies against mannan from Saccharomyces cerevisiae in human serum or plasma.
Detection Limit 0.5U/mL
Detection Method Colorimetric
Includes 1.5mL Calibrator with IgG and IgA class ASCA 6 vials, 1.5mL Each ASCA controls 2 vials, 20mL Sample Buffer 1 vial, 15mL Enzyme Conjugate solution 1 vial, 15mL Enzyme Conjugate solution 1 vial, 15mL TMB Substrate solution 1 vial, 15mL stop solution 1 vial, 96-well plate 1 plate, 20mL Wash solution 1 vial
Quantity 1 Kit
Sample Type Plasma, Serum
Sample Volume 100 μL
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type ASCA IgG/IgA ELISA kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.