Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ ATF2 (phospho T69/T71) Transcription Factor Assay

Catalog No. 89024588 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-024-588 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-024-588 Supplier Abnova™ Supplier No. KA1298

ATF2 (phospho T69/T71) Transcription Factor Assay is a colorimetric method for determining ATF2 DNA binding activity

Functional Study

Specifications

Immunoassay Kit Format Colorimetric Detection Kit
Product Type ATF2 (Phospho T69/T71) transcription facto r assay kit
Sufficient For 15 Reactions
For Use With (Application) Functional study, for determining ATF2 DNA binding activity
Quantity Each
Storage Requirements -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.