Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ BCP Albumin Assay Kit

Catalog No. 89028345 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-345 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-345 Supplier Abnova™ Supplier No. KA1613
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • Reagent, 50mL
  • Albumin standard, 2mL 5g/dL BSA

BCP Albumin Assay Kit is a quantitative colorimetric albumin determination at 610 nm.

  • Sensitive and accurate: Use as little as 20 μL samples
  • Simple and high throughput: procedure involves addition of a single working reagent and incubation for 5 minutes
  • Can be readily automated as a high-throughput assay for thousands of samples per day
  • Improved reagent stability and versatility: The optimized formulation has greatly enhanced reagent and signal stability
  • cuvette or 96-well plate assay
  • Low interference in biological samples, no pretreatments needed, and can be performed directly on raw biological samples
  • Store reagent and standard at 4°C and -20°C, respectively; see expiry dates on labels

Specifications

Content And Storage Store Reagent at 4°C. Store Standard at -20°C.
Description BCP Albumin Assay Kit is a quantitative colorimetric albumin determination at 610 nm.
Product Type BCP Albumin Assay Kit
For Use With (Application) Functional Study
Kit Contents 50mL Reagent, Albumin standard, 2mL 5g/dL BSA
Quantity Each
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.