Learn More
Description
- Simple labeling process
- Contains all of the necessary reagents for labeling including the storage buffer
- Five to eight biotin molecules conjugate to each IgG molecule on average
- Excess biotin molecules can be removed using a filtration tube included in this kit
Specifications
Specifications
| Description | Biotin Labeling Kit-NH2 is used for simple and rapid preparation of biotin-labeled antibodies and proteins. |
| For Use With (Application) | Conjugation |
| Product Type | Biotin Labeling Kit-NH2 |
| Quantity | 1 Kit |
| Content And Storage | NH2-Reactive Biotin (3 Tubes), 4 mL x 1 WS Buffer, 500 μL x 1 Reaction Buffer, Filtration Tube (3 Tubes). Store at 4°C. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.