Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Bisphenol A ELISA Kit

Catalog No. 89028335 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-335 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-335 Supplier Abnova™ Supplier No. KA1495
Add to Cart
Add to Cart

Competitive binding Enzyme Immunoassay for the quantitative measurement of Bisphenol A

Specifications

Calibration Range 10 to 1000ng/mL
Conjugate HRP
Description Bisphenol A ELISA Kit is a competitive binding enzyme immunoassay for the quantitative measurement of Bisphenol A.
Detection Limit 10ng/mL
Detection Method Colorimetric
Includes 8-well break away strip coated with BPA, 1.5mL/each vials within a plastic frame 1, ST 0-5-BPA standard 0, 0.01, 0.05, 0.2, 1, 5μg/mL, 0.10mL 251x Anti-BPA Polyclonal chicken Antibody, 0.15mL 101x Anti-IgY-Px Enzyme Conjugate, 125mL Dilu
Quantity 1 Kit
Sample Type Soil, Water
Sample Volume 100 μL
Content And Storage Store at 4°C on dry atmosphere away from light.
For Use With (Application) Quantification
Product Type Bisphenol A ELISA kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.