Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Blood/Cultured Cell Total RNA Purification Kit

Catalog No. 89028631 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-631 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-631 Supplier Abnova™ Supplier No. U0064
Add to Cart
Add to Cart

Blood/Cultured Cell Total RNA Purification Kit is designed specifically for purifying total RNA from fresh whole human blood and cultured animal cells.

  • Kit was designed specifically for purifying total RNA from fresh whole human blood and cultured cells
  • Entire procedure can be completed within 30 minutes
  • Applications include RT-PCR, Northern Blotting, Primer Extension, mRNA Selection, and cDNA Synthesis
  • Yield of 5-30 μg

Specifications

Content And Storage Store at room temperature.
Description Blood/Cultured Cell Total RNA Purification Kit is designed specifically for purifying total RNA from fresh whole human blood and cultured animal cells.
For Use With (Application) Purification
Includes 200mL RBC Lysis Buffer, 60mL RB Buffer, 30mL RT Buffer, 50mL W1 Buffer, 25mL Wash Buffer (add 100mL Ethanol), 6mL RNase-free Water, RB column, 2mL collection tube
Product Type Blood/Cultured Cell Total RNA Purification Kit
Quantity 1 Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.