Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Bordetella pertussis IgA/IgM ELISA Kit

Catalog No. 89028822 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
96 Assays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-822 96 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-822 Supplier Abnova™ Supplier No. KA2091
Add to Cart
Add to Cart

ELISA for the detection of specific IgA and/or IgM antibodies to Bordetella pertussis in Human Serum

Specifications

Conjugate HRP
Cross Reactivity Human
Description Bordetella pertussis IgA/IgM ELISA Kit is an enzyme linked immunosorbent assay for the detection of specific IgA and/or IgM antibodies to Bordetella pertussis in human serum.
Detection Method Colorimetric
Includes B. pertussis Antigen-coated microtiter plate 96 break apart well (8x12), 100mL 20X concentrated Wash Buffer (20X) 1 bottle, 30mL Serum Diluent 1 bottle, 40mL Conjugate Diluent 1 bottle, 0.75mL cut Off control IgA 1 vial, 0.75mL cut Off control IgM 1 vial,, 0.75mL P10 calibrator 1 vial, 0.75mL P50 calibrator 1vvial, 0.75mL P100 calibrator 1 vial, 0.2mL HRP Conjugate (300X) 1 vial, 14mL TMB-Substrate 1 bottle, 15mL stop solution 1 bottle, plate cover 1 unit
Quantity 96 Assays
Sample Type Serum
Sample Volume 50 μL
Content And Storage Store the kit at 4°C. Do not freeze.
For Use With (Application) Quantification
Product Type Bordetella pertussis IgA/IgM ELISA Kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.