Learn More
Specifications
Specifications
| Conjugate | HRP |
| Cross Reactivity | Human |
| Description | Bordetella pertussis IgG ELISA Kit is an enzyme linked immunosorbent assay for the semi-quantitative determination of specific IgG antibodies to Bordetella pertussis in human serum. |
| Detection Method | Colorimetric |
| Includes | B. pertussis Antigen-coated microtiter plate 96 break apart well (8x12), 100mL 20X concentrated Wash Buffer 1 bottle, 30mL Serum Diluent 1 bottle, 40mL Conjugate Diluent 1 Bottle, 2mL negative control 1 vial, 2mL positive control 1 vial, 0.75mL P10 calibrator 1 vial, 0.75mL P50 calibrator 1vvial, 0.75mL P100 calibrator 1 vial, 0.2mL HRP Conjugate (300X) 1 vial, 14mL TMB-Substrate 1 bottle, 15mL stop solution 1 bottle, plate cover 1 unit |
| Quantity | 96 Assays |
| Sample Type | Serum |
| Sample Volume | 50 μL |
| Content And Storage | Store the kit at 4°C. Do not freeze. |
| For Use With (Application) | Quantification |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.