Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Borrelia IgG ELISA Kit

Catalog No. 89028306 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-306 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-306 Supplier Abnova™ Supplier No. KA1466
Add to Cart
Add to Cart

Solid phase Enzyme Immunoassay for the quantitative measurement of IgG class autoantibodies against Borrelia

Specifications

Conjugate HRP
Description Borrelia IgG ELISA Kit is a solid phase enzyme immunoassay for the quantitative measurement of IgG class autoantibodies against Borrelia.
Detection Limit 4.5AU/mL
Detection Method Colorimetric
Includes 96-well ELISA break away strip coated with specific Antigen 1.3mL/each vial standard A to E, 0.2mL Anti-Human IgG Antibodies labelled with HRP, 125mL Wash Buffer concentrate, 125mL Dilution Buffer, 15mL chromogenic Substrate, 30mL stop solution
Quantity 1 Kit
Sample Type Cerebrospinal Fluid, Serum, Synovial Fluid
Sample Volume 100 μL
Content And Storage Store at 4°C on dry atmosphere away from light.
For Use With (Application) Quantification
Product Type Borrelia IgG ELISA kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.