Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ C-Peptide ELISA Kit

Catalog No. 89025098 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
96 Assays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-025-098 96 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-025-098 Supplier Abnova™ Supplier No. KA1259
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • Microtiterwells, 12 x 8 (break apart) strips, 96 wells; Wells coated with anti-mouse-antibody 96 plate
  • Standard (Standard 0-5) Concentrations: 0-16ng/mL (see exact value on the QC-Datasheet). The standards are calibrated against WHO approved Reference material IRR C-Peptide, code 84/510. Contain non-mercury preservative. 6 vials, lyophilized, 0.75mL
  • Sample Diluent, ready to use, Contains non-mercury preservative 1 vial, 3mL
  • Antiserum, Ready to use. Monoclonal mouse anti C-Peptide antibody contains non-mercury preservative. 1 vial, 7mL
  • Enzyme Conjugate, Ready to use. Biotinylated C-Peptide contains non-mercury preservative. 1 vial, 14mL
  • Enzyme Complex, Ready to use. Contains horseradish Peroxidase. Contains non-mercury preservative. 1 vial, 14ml
  • Substrate Solution, Ready to use. TMB 1 vial, 14mL

Indirect Solid Phase ELISA for the quantitative in-vitro diagnostic measurement of C-Peptide in Serum, Plasma and Urine

Specifications

Calibration Range 0.06 to 16ng/mL
Conjugate HRP
Cross Reactivity Human
Description C-Peptide ELISA Kit is a solid phase enzyme-linked immunosorbent assay for the quantitative in vitro diagnostic measurement of C-Peptide in serum, plasma and urine.
Detection Method Colorimetric
Includes 6 Vials lyophilized 0.75mL, standard (standard 0 to 5), 1 vial 3mL sample diluent, 1 vial 7mL AntiSerum, 1 vial 14mL Enzyme Conjugate, 1 vial 14mL Enzyme complex, 1 vial 14mL Substrate solution, 1 vial 14mL stop solution, 1 plate 96-well plate, 1 vial 30mL wash solution
Quantity 96 Assays
Sample Type Plasma, Serum, Urine
Sample Volume 100 μL
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type C-Peptide ELISA Kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.