Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ C1q Ab ELISA Kit

Catalog No. 89024566 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-024-566 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-024-566 Supplier Abnova™ Supplier No. KA1274
Add to Cart
Add to Cart

Includes

  • Divisible microplate consisting of 12 modules of 8 wells each, coated with highly purified C1q. Ready to use, Qty.1
  • Combined calibrators with IgG class Anti-C1q (A-F) in a serum/buffer matrix (PBS, BSA, NaN3 <0.1% (w/w)) containing: IgG: 0; 6.3; 12.5; 25; 50; and 100 U/mL. Ready to use, 6 vials, 1.5mL each
  • Anti-C1q controls in a serum/buffer matrix (PBS, BSA, NaN3 <0.1% (w/w)) positive (1) and negative (2), for the respective concentrations see the enclosed package insert. Ready to use, 2 vials, 1, 5mL each
  • Sample buffer (Tris, NaN3 <0,1% (w/w)), yellow, concentrate (5x) 1 vial, 20mL
  • Enzyme conjugate solution (PBS, PROCLIN 300 <0,5% (v/v)), (light red) containing polyclonal rabbit anti-human IgG; labelled with horseradish peroxidase. Ready to use, 1 vial, 15mL
  • TMB substrate solution. Ready to use, 1 vial, 15mL

Indirect solid phase Enzyme Immunoassay for the quantitative measurement of IgG class autoantibodies against C1q in Human Serum or Plasma

Specifications

Conjugate HRP
Cross Reactivity Human
Description C1q Ab ELISA Kit is an indirect solid phase enzyme immunoassay for the quantitative measurement of IgG class autoantibodies against C1q in human serum or plasma.
Detection Limit 0.5U/mL
Detection Method Colorimetric
Includes 1.5mL Combined calibrator with IgG class Anti-C1q 6 vials, 1.5mL Each Anti-C1q controls in a Serum/Buffer Matrix 2 vials, 20mL Sample Buffer 1 vial, 15mL Enzyme Conjugate solution 1 vial, 15mL TMB Substrate solution 1 vial, 15mL stop solution 1 vial, 96-well plate 1 plate, 20mL wash solution 1 vial
Quantity 1 Kit
Sample Type Plasma, Serum
Sample Volume 100 μL
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type C1Q Ab ELISA kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.