Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ C3 (Human) ELISA Kit

Catalog No. 89022893 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Type:
CCS, CSF, Milk, Saliva, Urine
Plasma, Serum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
89-022-893 Plasma, Serum
89-100-338 CCS, CSF, Milk, Saliva, Urine
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-022-893 Supplier Abnova™ Supplier No. KA0466
Add to Cart
Add to Cart

Includes

  • Human Complement C3 Microplate: A 96-well polystyrene microplate (12 strips of 8 wells) coated with a polyclonal antibody against human complement C3
  • Sealing Tapes: Each kit contains 3 pre-cut, pressure-sensitive sealing tapes that can be cut to fit the format of the individual assay
  • Human Complement C3 Standard: Human Complement C3 in a buffered protein base (60μg, lyophilized)
  • Biotinylated Complement C3: 1 vial, lyophilized
  • EIA Diluent Concentrate (10x): A 10-fold concentrated buffered protein base (30mL)
  • Wash Buffer Concentrate (20x): A 20-fold concentrated buffered surfactant (30mL)
  • Streptavidin-Peroxidase Conjugate (SP Conjugate): A 100-fold concentrate (μL)
  • Chromogen Substrate: A ready-to-use stabilized peroxidase chromogen substrate tetramethylbenzidine (8mL)
  • 9. Stop Solution: A 0.5 N hydrochloric acid to stop the chromogen substrate reaction (12mL)

Sandwich Enzyme Immunoassay for the quantitative measurement of Human C3

Specifications

Calibration Range 0.47 to 30μL/mL
Conjugate Biotin
Cross Reactivity Human
Description C3 (Human) ELISA Kit is a competitive enzyme immunoassay for the quantitative measurement of human C3.
Detection Limit 0.4μg/mL
Detection Method Colorimetric
Includes 96-well Antibody coated microwell, 60μg Human complement C3 standard, 30mL (10X) Biotinylated complement C3 1 vial, 30mL EIA diluent concentrate, 30mL Wash Buffer 20X, Streptavidin peroxidase Conjugate (SP Conjugate) 80, 8mL Chromogen Su
Quantity 1 Kit
Sample Type Plasma, Serum
Sample Volume 25 μL
Content And Storage Store components of the kit at 4°C or -20°C as described in the protocol.
Form Liquid
For Use With (Application) Quantification
Product Type C3 (Human) ELISA Kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.