Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ C4 (Human) ELISA Kit

Catalog No. 89028169 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-169 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-169 Supplier Abnova™ Supplier No. KA1021
Add to Cart
Add to Cart

Sandwich Enzyme Immunoassay for the quantitative measurement of Human complement C4 in Urine, Saliva and cell culture supernatants

Specifications

Calibration Range 0.078 to 20ng/mL
Conjugate HRP
Cross Reactivity Human
Description C4 (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human complement C4 in urine, saliva, and cell culture supernatants.
Detection Limit 0.075ng/mL
Detection Method Colorimetric
Includes 96-well Antibody coated microwell, 1.6μg Human complement C4 standard, 100X Biotinylated Complement C4 Antibody 1 vial, 30mL 10X MIX diluent concentrate, Wash Buffer concentrate 2 Streptavidin Peroxidase Conjugate (SP Conjugate) 80, 8mL
Quantity 1 Kit
Sample Type Cell Culture Supernatant, Milk, Saliva, Urine
Sample Volume 50 μL
Content And Storage Store SP Conjugate, biotinylated antibody and reconstituted standard at -20°C. Store all other components at 4°C. Avoid repeated freezing and thawing.
Form Liquid
For Use With (Application) Quantification
Product Type C4 (Human) ELISA kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.