Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ C7 (Human) ELISA Kit

Catalog No. 89100373 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-373 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-373 Supplier Abnova™ Supplier No. KA2115
Add to Cart
Add to Cart

Includes

  • Human Complement C7 Microplate: A polystyrene microplate coated with a polyclonal antibody against human C7, 6 (8 x 12) wells
  • Sealing Tapes
  • Human Complement C7 Standard: Human C7 in a buffered protein base (lyophilized), 160ng
  • Biotinylated Human Complement C7 Antibody (50x): A 50-fold biotinylated polyclonal antibody against human C7, 140μL
  • MIX Diluent Concentrate (10x)
  • Wash Buffer Concentrate (20x)
  • Streptavidin-Peroxidase Conjugate (SP Conjugate): A 100-fold concentrate
  • Chromogen Substrate: A ready-to-use stabilized peroxidase chromogen substrate tetramethylbenzidine
  • Stop Solution: A 0.5N hydrochloric acid

Use to measure quantitatively human C7. Sandwich enzyme immunoassay.

This assay employs a quantitative sandwich enzyme immunoassay technique that measures C7 in less than 4 hours

Specifications

Calibration Range 0.625 to 40ng/mL
Conjugate Biotin
Cross Reactivity Human, Monkey
Description C7 (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human C7.
Detection Limit 0.6ng/mL
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Cell Culture Supernatant, CSF, Milk, Plasma, Saliva, Serum and Urine
Sample Volume 50 μL
Sensitivity ∼0.6 ng/mL
Packaging 96-well
Content And Storage Store components of the kit at 4°C or -20°C as described in the protocol.
For Use With (Application) Quantification
Product Type C7 (Human) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.