Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ C8 (Human) ELISA Kit

Catalog No. 89101110 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-101-110 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-101-110 Supplier Abnova™ Supplier No. KA3814
Add to Cart
Add to Cart

Includes

  • C8 Microplate: A 96-well polystyrene microplate coated with a polyclonal antibody against C8, 96 wells
  • Sealing Tapes
  • C8 Standard: Human C8 in a buffered protein base (lyophilized), 40ng
  • Biotinylated C8 Antibody (50x): A 50-fold concentrated biotinylated polyclonal antibody against C8, 140μL
  • EIA Diluent Concentrate (10x)
  • Wash Buffer Concentrate (20x)
  • Streptavidin-Peroxidase Conjugate (SP Conjugate): A 100-fold concentrate, 80μL
  • Chromogen Substrate: A ready-to-use stabilized peroxidase chromogen substrate tetramethylbenzidine
  • Stop Solution: A 0.5N hydrochloric acid

Use to measure quantitatively human C8. Sandwich enzyme immunoassay.

This assay employs a quantitative sandwich enzyme immunoassay technique that measures C8 in less than 4 hours

Specifications

Conjugate Biotin
Cross Reactivity Human
Description C8 (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human C8.
Detection Limit 0.6ng/mL
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Cell Culture, Milk, Plasma, Saliva, Serum
Sample Volume 50 μL
Packaging 96-well
Content And Storage Store the kit at 4°C. For long term storage, please store at -20°C.
For Use With (Application) Quantification
Product Type C8 (Human) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.