Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ cAMP ELISA Kit

Catalog No. 89100925 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-925 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-925 Supplier Abnova™ Supplier No. KA3388
Add to Cart
Add to Cart

Includes

  • Coated Clear 96 Well Plate
  • Cyclic AMP Standard: Cyclic AMP at 1,500pmol/mL in a special stabilizing solution
  • Cyclic AMP Antibody: A sheep antibody specific for cyclic AMP
  • Cyclic AMP Conjugate
  • Sample Diluent Concentrate. CAUSTIC
  • Plate Primer: A neutralizing solution containing special stabilizers and additives
  • Acetic Anhydride: WARNING: Corrosive Lachrymator
  • Triethylamine: WARNING: Corrosive Lachrymator
  • Wash Buffer Concentrate: A 20X concentrate
  • TMB Substrate
  • Stop Solution: A 1M solution of hydrochloric acid. CAUSTIC
  • Plate Sealer

Use to detect quantitatively of cAMP in lysed cells, EDTA and heparin plasma, urine, saliva and tissue culture media samples. Enzyme-linked immunosorbent assay.

The kit is unique in that all samples and standards are diluted into an acidic Sample Diluent, which contains special additives and stabilizers, for cAMP measurement

Specifications

Calibration Range 0 to 150pmol/mL
Conjugate HRP
Description cAMP ELISA Kit is an enzyme-linked immunosorbent assay for the quantitative detection of cAMP in lysed cells, EDTA and heparin plasma, urine, saliva and tissue culture media samples.
Detection Limit 0.078pmol/mL
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Cell Lysate, Plasma (EDTA, Heparin), Saliva, Tissue Culture Medium, Urine
Sample Volume 50 μL
Sensitivity 0.64 pmol/mL
Packaging 96-well
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type cAMP ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.