Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Cancer Antigen 125 (Human) ELISA Kit

Catalog No. 89022684 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-022-684 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-022-684 Supplier Abnova™ Supplier No. KA0205
Add to Cart
Add to Cart

Sandwich enzyme immunoassay for the quantitative measurement of cancer antigen 125

Specifications

Antigen Cancer Antigen 125
Calibration Range 31.2 to 2000pg/mL
Conjugate HRP
Cross Reactivity Human
Description CA125 (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of cancer antigen 125.
Detection Limit 5U/mL
Detection Method Colorimetric
Includes Murine Monoclonal Anti-CA125 coated microtiter plate 96-well, 13mL Enzyme Conjugate reagent, 1mL each CA125 reference standards containing 0, 15, 50, 100, 200 and 400 U/mL of CA125, 50mL Wash Buffer 20X, 11mL TMB reagent (One-Step), 11mL 1N HCl stop solution
Quantity 1 Kit
Sample Type Serum
Sample Volume 100 μL
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type CA125 (Human) ELISA Kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.