Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Cardiolipin IgA ELISA Kit

Catalog No. 89027234 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-027-234 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-027-234 Supplier Abnova™ Supplier No. KA1096
Add to Cart
Add to Cart

Indirect solid phase ELISA for the quantitative measurement of IgA class autoantibodies against Cardiolipin in Human Serum or Plasma

Specifications

Calibration Range 7.5 to 120U/mL
Conjugate HRP
Description Cardiolipin Ab IgA ELISA Kit is an indirect solid phase ELISA for the quantitative measurement of IgA class autoantibodies against cardiolipin in human serum or plasma.
Detection Limit 1U/mL
Detection Method Colorimetric
Includes calibrator with IgA class, 1.5mL each Anti-cardiolipin Antibodies 6 vials, 1.5mL each Anti-cardiolipin controls 2 vials, 20mL Sample Buffer 1 vial, 15mL Enzyme Conjugate solution 1 vial, 15mL TMB Substrate solution 1 vial, 15mL stop solution 1 vial, 96-well plate 1 plate, 20mL wash solution 1 vial
Quantity 96 Tests
Sample Type Plasma, Serum
Sample Volume 10 μL
Content And Storage Store the kit at 4°C
Form Liquid
For Use With (Application) Quantification
Product Type Cardiolipin IgA ELISA Kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.