Learn More
Specifications
Specifications
| Calibration Range | Not Applicable |
| Conjugate | HRP |
| Description | Cardiolipin IgG ELISA Kit is used for the detection of IgG antibody to Cardiolipin in human serum or plasma. |
| Detection Method | Colorimetric |
| Includes | Microwell coated with Cardiolipin Antigen 12x8x1, 22mL sample diluent,1mL calibrator 1 vial, 1mL positive control 1 vial, 1mL negative control 1 vial, 12mL Enzyme Conjugate 1 bottle, 12mL TMB Substrate, 12mL stop solution,20X 25mL wash concentrate |
| Quantity | 96 Assays |
| Sample Type | Plasma, Serum |
| Sample Volume | 10 μL |
| Content And Storage | Store the kit at 4°C |
| For Use With (Application) | Qualitative |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.