Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ CCL2 (Mouse) ELISA Kit

Catalog No. 89100786 Shop All Abnova Corporation Products
Catalog No. 89-100-786 Supplier Abnova™ Supplier No. KA3083
Add to Cart
Add to Cart

Includes

  • One 96-well plate pre-coated with anti-Mouse MCP-1 antibody, 96 (8 x 12) wells
  • Lyophilized MCP-1 standards, 10ng x 2
  • Sample/Standard diluent buffer, 30mL
  • Biotin conjugated anti-Mouse MCP-1 antibody (Concentrated), Dilution: 1:100, 130μL
  • Antibody diluent buffer, 12mL
  • Avidin-Biotin-Peroxidase Complex (ABC) (Concentrated), Dilution: 1:100, 130μL
  • ABC diluent buffer, 12mL
  • TMB substrate, 10mL
  • Stop solution, 10mL
  • Wash buffer (25X), 30mL

Use to measure quantitatively mouse Ccl2.

This kit is based on sandwich enzyme-linked immune-sorbent assay technology

Specifications

Calibration Range 15.6 to 1000pg/mL
Conjugate Biotin
Cross Reactivity Mouse
Description Ccl2 (Mouse) ELISA Kit is intended for the quantitative measurement of mouse Ccl2.
Detection Limit 6pg/mL
Detection Method Colorimetric
Includes One 96-well plate pre-coated with anti-Mouse MCP-1 antibody (96 (8 x 12) wells), Lyophilized MCP-1 standards (10ng x 2), Sample/Standard diluent buffer (30mL), Biotin conjugated anti-Mouse MCP-1 antibody (Concentrated),Dilution: 1:100 (130μL), Antibody diluent buffer (12mL), Avidin-Biotin-Peroxidase Complex (ABC) (Concentrated), Dilution: 1:100 (130μL), ABC diluent buffer (12mL), TMB substrate (10mL), Stop solution (10mL), Wash buffer 25X (30mL)
Regulatory Status For research use only (RUO)
Sample Type Body Fluid, Cell Culture Supernatant, Plasma, Serum, Tissue Lysate
Sample Volume 100 μL
Sensitivity <6 pg/mL
Packaging 96-well
Content And Storage Store at 4°C for 4 months, or at -20°C for 8 months.
For Use With (Application) Quantification
Product Type Ccl2 (Mouse) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.