Learn More
Description
- Ccl20 (Rat) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of rat Ccl20.
- Target Protein: Ccl20
- Reactivity: Rat
- Application: Quantitative measurement
- Calibration Range: 7.8 to 500 pg/mL
- Regulatory Status: For research use only (RUO)
- Storage: Store at 4 °C for 6 months, at -20 ° C for 12 months.
- Avoid multiple freeze-thaw cycles.
Specifications
Specifications
| Calibration Range | 7.8 to 500pg/mL |
| Conjugate | Biotin |
| Cross Reactivity | Rat |
| Description | Ccl20 (Rat) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of rat Ccl20. |
| Detection Limit | <1pg/mL |
| Detection Method | Colorimetric |
| Includes | 96-well Antibody coated microwell, 10ng x 2 vials lyophilized recombinant Rat Ccl20 standard, 30mL Sample Diluent Buffer, 130μL Biotinylated Anti-Rat Ccl20 Antibody,, 12mL Antibody Diluent Buffer,130μL avidin biotin per |
| Quantity | 1 Kit |
| Sample Type | Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum |
| Sample Volume | 100 μL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.