Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ CCL21 (Human) ELISA Kit

Catalog No. 89101012 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-101-012 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-101-012 Supplier Abnova™ Supplier No. KA3333
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • 96-well plate precoated with anti-human CCL21 antibody 96 (8 x 12) wells
  • Lyophilized recombinant human CCL21 standard 10ng x 2
  • Biotinylated anti-human CCL21 antibody(dilution 1:100) 130μL
  • Avidin-Biotin-Peroxidase Complex (ABC) (dilution 1:100) 130μL
  • Sample diluent buffer 30mL
  • Antibody diluent buffer 12mL
  • ABC diluent buffer 12mL
  • TMB color developing agent 10mL
  • TMB stop solution 10mL

Use to measure quantitatively human CCL21. Sandwich enzyme immunoassay.

CCL21 (Human) ELISA Kit is based on standard sandwich enzyme-linked immune-sorbent assay technology

Specifications

Calibration Range 62.5 to 4000pg/mL
Conjugate Biotin
Cross Reactivity Human
Description CCL21 (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human CCL21.
Detection Limit 12pg/mL
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status RUO
Sample Type Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum
Sample Volume 100 μL
Sensitivity <12 pg/mL
Packaging 96-well
Content And Storage Store the kit at -20°C
For Use With (Application) Quantification
Product Type CCL21 (Human) ELISA Kit
Specificity Natural and recombinant human CCL21
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.