Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ CCL23 (Human) ELISA Kit

Catalog No. 89028486 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
96 Assays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-486 96 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-486 Supplier Abnova™ Supplier No. KA1751

missing translation for 'validMainframeMsg'

Includes

  • MPIF-1 Microplate (Item A): 96 wells (12 strips x 8 wells) coated with anti-human MPIF-1, 96-wells (12 strips x 8 wells)
  • Wash Buffer Concentrate (20x) (Item B), 25mL
  • Standards (Item C), recombinant human MPIF-1, 2 vials
  • Assay Diluent A (Item D): animal serum with 0.09% sodium azide as preservative. For Standard/Sample (serum/plasma) diluent, 30mL
  • Assay Diluent B (Item E): 5x concentrated buffer, For Standard/Sample (cell culture medium/urine) diluent, 15mL
  • Detection Antibody MPIF-1 (Item F): biotinylated anti-human MPIF-1 (each vial is enough to assay half microplate), 2 vial
  • HRP-Streptavidin Concentrate (Item G): 200x concentrated HRP-conjugated streptavidin, 200μL
  • TMB One-Step Substrate Reagent (Item H): 3, 3', 5, 5'- tetramethylbenzidine (TMB) in buffered solution, 12mL
  • Stop Solution (Item I): 0.2M sulfuric acid, 8mL

In-Vitro Enzyme-Linked Immunosorbent assay for the quantitative measurement of Human CCL23

Specifications

Conjugate Biotin
Cross Reactivity Human
Description CCL23 (Human) ELISA Kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human CCL23.
Detection Limit 7pg/mL
Detection Method Colorimetric
Includes 96-well Antibody coated microwell, 25mL Wash Buffer 20X, 2 vials standard, 30mL assay diluent A, 15mL assay diluent B,, 2 vials detection Antibody MSP,8μL HRP Streptavidian concentrrate,12mL TMB one step Substrate reagent,8mL stop soluti
Quantity 96 Assays
Sample Type Cell Culture Supernatant, Plasma, Serum, Urine
Sample Volume 100 μL
Content And Storage Store at 4°C. Standard (Recombinant protein) should be stored at -80°C after reconstitution. The whole kit can be stored at -20°C for long term storage. Avoid repeated freezing and thawing.
For Use With (Application) Quantification
Product Type CCL23 (Human) ELISA Kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.