Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ CCL4 (Human), Biotin, ELISA Kit

Catalog No. 89101011 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-101-011 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-101-011 Supplier Abnova™ Supplier No. KA3332
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • 96-well plate precoated with anti- human CCL4 antibody 96 (8x12) wells
  • Lyophilized recombinant human CCL4 standard(dilution 1:100) 10ng x 2
  • Biotinylated anti- human CCL4 antibody(dilution 1:100) 130μL
  • Avidin-Biotin-Peroxidase Complex (ABC) 130μL
  • Sample diluent buffer 30mL
  • antibody diluent buffer 12mL
  • ABC diluent buffer 12mL
  • TMB color developing agent 10mL
  • TMB stop solution 10mL

Sandwich Enzyme Immunoassay for the quantitative measurement of Mouse Igfbp3

Specifications

Calibration Range 15.6 to 1000pg/mL
Conjugate Biotin
Cross Reactivity Human
Description CCL4 (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human CCL4.
Detection Limit 1 pg/mL
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Cell culture supernate, plasma (Citrate, EDTA, Heparin) and serum
Sample Volume 100 μL
Content And Storage Store the kit at -20°C
Form Lyophilized
For Use With (Application) Quantification
Product Type CCL4 (Human) ELISA Kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.