Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ CCL7 (Human) ELISA Kit

Catalog No. 89100208 Shop All Abnova Corporation Products
Change view
Click to view available options
Calibration Range:
15 to 800 pg/mL
15.6 to 1000 pg/mL
30 to 1600 pg/mL
Detection Limit:
1 pg/mL
10 pg/mL
30 pg/mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Calibration Range Detection Limit
89-100-208 30 to 1600 pg/mL 30 pg/mL
89-100-788 15 to 800 pg/mL 10 pg/mL
89-101-008 15.6 to 1000 pg/mL 1 pg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 89-100-208 Supplier Abnova™ Supplier No. KA0519
Add to Cart
Add to Cart

Includes

  • MCP-3 Microtiter plate, 96 wells
  • MCP-3 Conjugate, 12mL
  • MCP-3 Standard, 2 vials
  • Calibrator diluent I, 25mL
  • Calibrator diluent II, 25mL
  • Wash Buffer (20x), 60mL
  • Substrate A, 10mL
  • Substrate B, 10mL
  • Stop solution, 14mL

This MCP-3 enzyme linked immunosorbent assay (ELISA) applies a technique called a quantitative sandwich immunoassay

Specifications

Calibration Range 30 to 1600 pg/mL
Conjugate HRP
Cross Reactivity Human
Description CCL7 (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human CCL7.
Detection Limit 30 pg/mL
Detection Method Colorimetric
Includes Antibody coated microwells (96 wells), MCP-3 CONJUGATE (15mL), MCP-3 STANDARD (6.4ng/vial × 2), Calibrator diluent I (22mL), Calibrator diluent II (22mL), Sample diluent (12mL), Wash buffer 20X (60mL), Substrate A and B (11mL each), Stop solution (14mL)
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type CCS, Plasma, Serum
Sample Volume 100 μL
Sensitivity With Calibrator Diluent I is 30 pg/mL, with Calibrator Diluent II is 20 pg/mL
Packaging 96-well
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type CCL7 (Human) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.