Learn More
Description
Includes
- Precoated ELISA modules, 1 plate
- Detecting antibody (POD-labelled monoclonal antibody to human CD14) Ready for use, 1 vial
- Human-CD14-standard (recombinant human CD14, lyophilized), 1 vial
- Reference serum (3.2 ± 0.6μg/mL, lyophilized), 1 vial
- PBS (Phosphate buffer salt solution), 2 tab
- Dilution Buffer, 1 vial
- Tween 20, 1 vial
- Stopping solution Ready for use, 1 vial
- Substrate solution Ready for use, 1 vial
Specifications
Specifications
| Calibration Range | 5 to 50ng/mL |
| Conjugate | POD |
| Cross Reactivity | Human |
| Description | CD14 (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human CD14. |
| Detection Method | Colorimetric |
| Quantity | 1 Kit |
| Regulatory Status | RUO |
| Sample Type | Culture Medium, Plasma, Serum |
| Sample Volume | 100 μL |
| Content And Storage | Store the kit at 4°C. Follow protocol instructions for long term storage. Avoid repeated freezing and thawing. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.