Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ CD14 (Mouse) ELISA Kit

Catalog No. 89100258 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-258 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-258 Supplier Abnova™ Supplier No. KA0447
Add to Cart
Add to Cart

Includes

  • Precoated ELISA modules, 96 wells
  • Detecting antibody (HRP-labelled polyclonal antibodies to mouse CD14), 1 vial
  • CD14-standard (recombinant mouse CD14, lyophilized), 1 vial
  • Reference serum (lyophilized), 1 vial
  • PBS, 2 tablets
  • Dilution buffer for samples, 1 vial
  • Tween 20, 1 vial
  • Stopping solution, Ready for use, 1 vial
  • Substrate solution , Ready for use, 1 vial
  • Dilution buffer for Detecting antibody, 1 vial

Use to measure quantitatively mouse Cd14. Sandwich enzyme immunoassay.

The Cd14 (Mouse) ELISA Kit is a solid phase sandwich Enzyme-Linked-Immunosorbent-Assay (ELISA)

Specifications

Calibration Range 5 to 50ng/mL
Conjugate POD
Cross Reactivity Mouse
Description Cd14 (Mouse) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of mouse Cd14.
Detection Limit 5ng/mL
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status RUO
Sample Type Culture Medium, Plasma, Serum
Sample Volume 50 μL
Packaging 96-well
Content And Storage Store at 4°C. For long term storage of CD14-standard and Reference serum store at -20°C or -80°C. Aliquot to avoid repeated freezing and thawing.
For Use With (Application) Quantification
Product Type Cd14 (Mouse) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.