Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ CDH2 (Human) ELISA Kit

Catalog No. 89027273 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
96 Assays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-027-273 96 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-027-273 Supplier Abnova™ Supplier No. KA1135

CDH2 (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human CDH2.

Quantification

Specifications

Calibration Range 312 to 20000pg/mL
Conjugate HRP
Cross Reactivity Human
Detection Limit 78pg/mL
Detection Method Colorimetric
Includes 1 Assay plate, standard 2, 20mL sample diluent, 10mL assay diluent A, 10mL assay diluent B, 1200μL detection reagent A, 1200μL detection reagent B, 30mL 25X Wash Buffer, 10mL Substrate,10mL stop solution,5 plate sealer
Quantity 96 Assays
Sample Type Biological Fluid, Cell Culture Supernatant, Plasma, Serum, Tissue Homogenate
Sample Volume 100 μL
Content And Storage 4°C
For Use With (Application) In-vitro quantitative determination of Human Cadherin-2 (CDH2) concentrations
Product Type CDH2 (Human) ELISA kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.