Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ CDH3 (Mouse) ELISA Kit

Catalog No. 89101032 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-101-032 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-101-032 Supplier Abnova™ Supplier No. KA3353
Add to Cart
Add to Cart

Use to measure quantitatively mouse Cdh3. Sandwich enzyme immunoassay.

Specifications

Calibration Range 62.5 to 4000pg/mL
Conjugate Biotin
Cross Reactivity Mouse
Description Cdh3 (Mouse) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of mouse Cdh3.
Detection Method Colorimetric
Quantity 1 Kit
Sample Type Cell Culture Supernatant, Serum
Sample Volume 100 μL
Content And Storage Store the kit at -20°C
For Use With (Application) Quantification
Product Type Cdh3 (Mouse) ELISA Kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.