Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ CEA (Human) ELISA Kit

Catalog No. 89100040 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-040 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-040 Supplier Abnova™ Supplier No. KA0204
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • Antibody-coated microtiter plate with 96 wells, 1
  • CEA standards containing; 0, 3, 12, 30, 60, and 120ng/mL CEA. Ready to use, 1mL x 6
  • Enzyme Conjugate Reagent, 13mL
  • TMB Reagent (One-Step), 11mL
  • Stop Solution (1N HCl), 11mL

Use to measure quantitatively carcinoembryonic antigen. Sandwich enzyme immunoassay.

The CEA (Human) ELISA Kit is based on the principle of a solid phase enzyme-linked immunosorbent assay

Specifications

Calibration Range 0 to 120ng/mL
Conjugate HRP
Cross Reactivity Human
Description CEA (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of carcinoembryonic antigen.
Detection Limit 1ng/mL
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status RUO
Sample Type Serum
Sample Volume 50 μL
Packaging 96-well
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type CEA (Human) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.