Learn More
Description
Comprend
FISH Hybridization Buffer (80μL)
FISH is a technique used to identify and localize the presence or absence of specific DNA sequences on cells and tissues. Abnova has developed a range of FISH probes for the detection of gene amplification, loss, and translocation. Each FISH probe has a pair of locus-specific, fluorophore-labeled probes originated from a bacterial chromosome (BAC) library.
Advantages
- Dual colored probes for fast, sensitive, and specific detection
- Work on metaphase spread, paraffin embedded, and frozen tissue
- Identify gene amplification, loss, and translocation
- High signal to noise ratio
- Low cross reactivity
Spécifications
Spécifications
| Content And Storage | Store at 4°C in the dark. |
| Label or Dye | Texas Red |
| For Use With (Application) | Fluorescent In Situ Hybridization (Cell) |
| Includes | FISH Hybridization Buffer (80 uL) |
| Location | 19q12 |
| Probe | CEN19q |
| Product Type | CEN19q (Texas Red) FISH Probe |
| Quantity | 20 μL |
missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.