Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ CFD (Mouse) ELISA Kit

Catalog No. 89101118 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-101-118 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-101-118 Supplier Abnova™ Supplier No. KA3822
Add to Cart
Add to Cart

Includes

  • Mouse Complement Factor D Microplate: A 96-well polystyrene microplate coated with a polyclonal antibody against mouse FD
  • Sealing Tapes
  • Mouse Complement Factor D Standard: Mouse FD in a buffered protein base (lyophilized)
  • Biotinylated Mouse Complement Factor D Antibody (50x)
  • EIA Diluent Concentrate (10x)
  • Wash Buffer Concentrate (20x): A 20-fold concentrated buffered surfactant
  • Streptavidin-Peroxidase Conjugate (SP Conjugate): A 100-fold concentrate
  • Chromogen Substrate: A ready-to-use stabilized peroxidase chromogen substrate tetramethylbenzidine
  • Stop Solution: A 0.5 N hydrochloric acid

Use to measure quantitatively mouse Cfd. Sandwich enzyme immunoassay.

This assay employs a quantitative sandwich enzyme immunoassay technique that measures mouse FD in less than 4 hours

Specifications

Calibration Range 0.031 to 1.0ng/mL
Conjugate Biotin
Cross Reactivity Mouse
Description Cfd (Mouse) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of mouse Cfd.
Detection Limit 0.01ng/mL
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Cell Culture, Plasma, Serum, Urine
Sample Volume 50 μL
Packaging 96-well
Content And Storage Store SP Conjugate and Biotinylated Antibody at -20°C. Store Standard at 2-8°C before reconstituting with Diluent and at -20°C after reconstituting with Diluent. Store all other components at 4°C.
For Use With (Application) Quantification
Product Type Cfd (Mouse) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.