Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ CHGA (Human) ELISA Kit

Catalog No. 89028617 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-617 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-617 Supplier Abnova™ Supplier No. KA1884
Add to Cart
Add to Cart

Enzyme Immunoassay for the quantitative determination of chromogranin A in Human Serum, Plasma

Specifications

Cross Reactivity Human
Description Chromogranin A ELISA Kit is an enzyme immunoassay for the quantitative determination of Chromogranin A in human serum, plasma.
Detection Limit 10μg/L
Detection Method Colorimetric
Includes 1 x 20mL Wash Buffer concentrate, 1 x 12mL Substrate, 1 x 12mL stop solution, 1 x 1mL standard A, 1 x 1mL standard B, 1 x 1mL standard C, 1 x 1mL standard D, 1 x 1mL standard E, 1 x 50mL Assay Buffer, 1 plate chromogranin A microtiter strip, 1 x 12mL Conjugate,1 x 12mL biotin Antibody,1 x 4mL adjustment Buffer,1 x 1mL control 1,1 x 1mL control 2
Quantity 1 Kit
Sample Type Plasma, Serum
Sample Volume 25 μL
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type CHGA (Human) ELISA kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.