Learn More
Specifications
Specifications
| Conjugate | HRP |
| Cross Reactivity | Chlamydia species |
| Description | Chlamydia Trachomatis IgM ELISA Kit is used for the detection of IgM antibody to Chlamydia Trachomatis in human serum or plasma. |
| Detection Method | Colorimetric |
| Includes | 12 x 8 x 1 Microwell, 22mL sample diluent, 1.5mL calibrator, 1.5mL positive control, 1.5mL negative control, 12mL Enzyme Conjugate, 12mL TMB chromogenic Substrate,, 12mL stop solution,25mL 20X washing concentrate |
| Quantity | 1 Kit |
| Sample Type | Plasma, Serum |
| Sample Volume | 10 μL |
| Content And Storage | Store the kit at 4°C |
| For Use With (Application) | Qualitative |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.