Learn More
Abnova™ Chromatin Immunoprecipitation (ChIP) Kit
Catalog No. 89027042 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
89-027-042 1 Kit
1 options
Catalog No. 89-027-042 Supplier Abnova™ Supplier No. KA0685
Add to Cart
Add to Cart
Chromatin Immunoprecipitation (ChIP) Kit is use for investigating protein-DNA interaction in vivo efficiently.
  • The fastest procedure available, which can be finished within 5 hours
  • Strip microplate format makes the assay flexible, manual or high throughput
  • Columns for DNA purification are included, which saves time and reduces labor
  • Compatible with all DNA amplification-based approaches
  • Simple, reliable, and consistent assay conditions
  • Store the following components at 4°C: Protease inhibitor cocktail, normal mouse lgG, Anti-RNA polymerase II, proteinase K, control primers, and 8-well assay strips; store all other components at room temperature
  • Stable for up to 6 months when stored properly
Specifications
Description Chromatin Immunoprecipitation (ChIP) Kit is use for investigating protein-DNA interaction in vivo efficiently.
Content And Storage Store the following components at 4°C: Protease inhibitor cocktail, normal mouse IgG, Anti-RNA polymerase II, proteinase K, control primers, 8 well strips. Store other components at room temperature.
Quantity 1 Kit
For Use With (Application) Chromatin Immunoprecipitation
Type ChIP Kit