Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Clostridium difficile GDH ELISA Kit

Catalog No. 89100986 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-986 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-986 Supplier Abnova™ Supplier No. KA3381

missing translation for 'validMainframeMsg'

Includes

  • Microtiter plate coated with anti-GDH monoclonal antibody
  • Concentrated Wash Buffer (20x): PBS -Tween buffer
  • Sample Diluent
  • HRP-Conjugate (green): A ready-to-use solution containing Horseradish peroxidase (HRP) conjugated anti-GDH monoclonal antibody. Contains less than 0.05% Proclin as preservative
  • Positive Control: A ready to use solution containing recombinant GDH protein. Contains less than 0.05% Proclin as preservative
  • TMB-Substrate
  • Stop Solution: A ready to use solution. Contains 1M H2SO4
  • Disposable plastic pipettes
  • Plate cover

For the qualitative detection of Clostridium difficile GDH in human fecal specimens. Enzyme-linked immunosorbent assay.

Clostridium difficile GDH ELISA Kit is an Enzyme-Linked Immunosorbent Assay (ELISA) for use as a screening test to detect Clostridium difficile glutamate dehydrogenase (GDH) in human fecal specimens from individuals suspected of having C. difficile disease

Specifications

Conjugate HRP
Cross Reactivity Human
Description Clostridium difficile GDH ELISA Kit is an enzyme linked immunosorbent assay for qualitative detection of Clostridium difficile GDH in human fecal specimens.
Detection Limit 1ng/well
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Feces
Sample Volume 0.05 to 0.1g
Packaging 96-well
Content And Storage Store the kit at 4°C. Do not freeze.
For Use With (Application) Qualitative
Product Type Clostridium difficile GDH ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.