Learn More
Description
Clu (Mouse) ELISA Kit is an enzyme immunoassay for the quantitative determination of Clu in mouse serum and plasma.
Quantification
Specifications
Specifications
| Conjugate | HRP |
| Description | Clu (Mouse) ELISA Kit is for the quantitative measurement of Clu in plasma and serum. |
| Quantity | 1 Kit |
| Content And Storage | Store the kit at 4°C |
| For Use With (Application) | Quantification |
| Product Type | Clu (Mouse) ELISA Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.