Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ COL18A1 (Human) ELISA Kit

Catalog No. 89101057 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Type:
CCS, Human Milk, Plasma (EDTA, Heparin), Serum
CCS, Plasma, Serum, Urine
Cell Lysates, Plasma, Serum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
89-101-057 CCS, Human Milk, Plasma (EDTA, Heparin), Serum
89-100-478 Cell Lysates, Plasma, Serum
89-028-444 CCS, Plasma, Serum, Urine
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 89-101-057 Supplier Abnova™ Supplier No. KA3409
Add to Cart
Add to Cart

In-Vitro Enzyme-Linked Immunosorbent assay for the quantitative measurement of Human COL18A1

Specifications

Calibration Range 156 to 10000pg/mL
Conjugate Biotin
Cross Reactivity Human
Description COL18A1 (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human COL18A1.
Detection Limit <10pg/mL
Detection Method Colorimetric
Includes 96-well plate pre-coated with anti-Human Endostatin antibody, Lyophilized Endostatin standards (10ng x 2), Sample diluent buffer (30mL), Biotin conjugated anti-Human Endostatin antibody (130uL), Antibody diluent buffer (12mL), Avidin-Biotin-Peroxidase Complex (130uL), Antibody diluent buffer (12mL), ABC diluent buffer (12mL), TMB substrate (10mL), Stop solution (10mL)
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type CCS, Human Milk, Plasma (EDTA, Heparin), Serum
Sample Volume 100 μL
Content And Storage Store the kit at -20°C
For Use With (Application) Quantification
Product Type COL18A1 (Human) ELISA kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.